2-Chloro-5-(trifluoromethyl)-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-5-(trifluoromethyl)benzo[d]oxazole

PC300516-250MG
APOSPC300516-250MGEA 53.8 EUR
APOSPC300516-250MG
2-Chloro-5-(trifluoromethyl)-1,3-benzoxazole
2-Chloro-5-(trifluoromethyl)-1,3-benzoxazole
Formel: C₈H₃ClF₃NO
Molecular Weight: 221.564 g/mol
Schmelzpunkt: 26…28 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18250484
CAS-Nummer: 114997-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR