N-Methyl-N-[2-morpholin-4-yl-5-(trifluoromethyl)benzyl]amine
Lieferant: Apollo Scientific
|
Gefahr
|
PC300519-1G
APOSPC300519-1GEA
145
EUR
APOSPC300519-1G
N-Methyl-N-[2-morpholin-4-yl-5-(trifluoromethyl)benzyl]amine
N-Methyl-N-[2-morpholin-4-yl-5-(trifluoromethyl)benzyl]amine
Formel:
C₁₃H₁₇F₃N₂O Molecular Weight: 274,282 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025891 CAS-Nummer: 886851-52-1 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...