1-(3-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Lieferant: Apollo Scientific
PC300524-1G
APOSPC300524-1GEA
147
EUR
APOSPC300524-1G
1-(3-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Formel:
C₁₁H₁₀FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569310 CAS-Nummer: 566154-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...