tert-Butyl 4-[3-(4-fluorophenoxy)-2-hydroxypropyl]piperazine-1-carboxylate

Lieferant: Apollo Scientific

PC300527-1G
APOSPC300527-1GEA 248 EUR
APOSPC300527-1G
tert-Butyl 4-[3-(4-fluorophenoxy)-2-hydroxypropyl]piperazine-1-carboxylate
tert-Butyl 4-[3-(4-fluorophenoxy)-2-hydroxypropyl]piperazine-1-carboxylate
Formel: C₁₈H₂₇FN₂O₄
Molecular Weight: 354,416 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664816
CAS-Nummer: 874801-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR