3-Chloro-4-(trifluoromethyl)-1H-indazole

Lieferant: Apollo Scientific
Achtung

PC300534-1G
APOSPC300534-1GEA 511 EUR
APOSPC300534-1G
3-Chloro-4-(trifluoromethyl)-1H-indazole
3-Chloro-4-(trifluoromethyl)-1H-indazole
Formel: C₈H₄ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22563699
CAS-Nummer: 1388070-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR