2-(4-Bromo-1H-pyrazol-1-yl)-3-chloro-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC300537-1G PC300537-5G PC300537-10G
APOSPC300537-1GEA 186 EUR
APOSPC300537-1G APOSPC300537-5G APOSPC300537-10G
2-(4-Bromo-1H-pyrazol-1-yl)-3-chloro-5-(trifluoromethyl)pyridine
2-(4-Bromo-1H-pyrazol-1-yl)-3-chloro-5-(trifluoromethyl)pyridine
Formel: C₉H₄BrClF₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12861968
CAS-Nummer: 1183812-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR