3-Chloro-2-phenoxy-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC300538-1G PC300538-5G
APOSPC300538-1GEA 0 EUR
APOSPC300538-1G APOSPC300538-5G
3-Chloro-2-phenoxy-5-(trifluoromethyl)pyridine
3-Chloro-2-phenoxy-5-(trifluoromethyl)pyridine
Formel: C₁₂H₇ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00244669
CAS-Nummer: 95711-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR