4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid

Lieferant: Apollo Scientific
Achtung

PC300540-1G PC300540-250MG
APOSPC300540-1GEA 0 EUR
APOSPC300540-1G APOSPC300540-250MG
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Formel: C₁₃H₇ClF₃NO₃
Schmelzpunkt: 156…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11840267
CAS-Nummer: 105626-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR