1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-sulphonyl chloride

Lieferant: Apollo Scientific

PC300543-1G
APOSPC300543-1GEA 361 EUR
APOSPC300543-1G
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-sulphonyl chloride
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-sulphonyl chloride
Formel: C₉H₄Cl₂F₃N₃O₂S
Molecular Weight: 346,113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13722967
CAS-Nummer: 1156603-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR