3-(Dimethylamino)-1-(4-fluorophenyl)-2-propen-1-one

Lieferant: Apollo Scientific

PC300544-5G
APOSPC300544-5GEA 369 EUR
APOSPC300544-5G
3-(Dimethylamino)-1-(4-fluorophenyl)-2-propen-1-one
3-(Dimethylamino)-1-(4-fluorophenyl)-2-propen-1-one
Formel: C₁₁H₁₂FNO
Molecular Weight: 193,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00111509
CAS-Nummer: 75175-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR