Ethyl-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Lieferant: Apollo Scientific
PC300546-1G
PC300546-5G
APOSPC300546-1GEA
139
EUR
APOSPC300546-1G
APOSPC300546-5G
Ethyl-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Ethyl-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formel:
C₁₂H₉F₃O₂S Molecular Weight: 274.26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100111 CAS-Nummer: 1002355-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...