7-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

PC300547-1G
APOSPC300547-1GEA 226 EUR
APOSPC300547-1G
7-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide
7-(Trifluoromethyl)-1-benzothiophene-2-carbohydrazide
Formel: C₁₀H₇F₃N₂OS
Molecular Weight: 260,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111408
CAS-Nummer: 1171927-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR