3{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanohydrazide

Lieferant: Apollo Scientific

PC300549-1G
APOSPC300549-1GEA 224 EUR
APOSPC300549-1G
3{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanohydrazide
3{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]thio}propanohydrazide
Formel: C₉H₉ClF₃N₃OS
Molecular Weight: 299.7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100119
CAS-Nummer: 1215738-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR