1-(2-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC300552-1G
APOSPC300552-1GEA 65.4 EUR
APOSPC300552-1G
1-(2-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(2-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₁H₁₀FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01330894
CAS-Nummer: 1998-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR