5-(Trifluoromethyl)quinazolin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
PC300553-1G
APOSPC300553-1GEA
447
EUR
APOSPC300553-1G
5-(Trifluoromethyl)quinazolin-2-amine
5-(Trifluoromethyl)quinazolin-2-amine
Formel:
C₉H₆F₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12406818 CAS-Nummer: 190273-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...