2'-Fluor-4'-nitroacetanilid

Lieferant: Apollo Scientific

Synonyme: N-(2-Fluoro-4-nitrophenyl)acetamide

PC300555-5G
APOSPC300555-5GEA 143 EUR
APOSPC300555-5G
2'-Fluor-4'-nitroacetanilid
2'-Fluor-4'-nitroacetanilid
Formel: C₈H₇FN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00033912
CAS-Nummer: 348-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR