N-Methyl-2-(trifluoromethyl)benzenesulphonamide

Lieferant: Apollo Scientific
Achtung

PC300559-1G
APOSPC300559-1GEA 0 EUR
APOSPC300559-1G
N-Methyl-2-(trifluoromethyl)benzenesulphonamide
N-Methyl-2-(trifluoromethyl)benzenesulphonamide
Formel: C₈H₈F₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD05270097
CAS-Nummer: 633697-56-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR