1-Methyl-6-(trifluoromethyl)-1H-indazol-3-amine

Lieferant: Apollo Scientific
Achtung

PC300562-1G
APOSPC300562-1GEA 364 EUR
APOSPC300562-1G
1-Methyl-6-(trifluoromethyl)-1H-indazol-3-amine
1-Methyl-6-(trifluoromethyl)-1H-indazol-3-amine
Formel: C₉H₈F₃N₃
Molecular Weight: 215,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847096
CAS-Nummer: 1260796-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR