N-Methyl-3-(trifluoromethyl)pyridin-2-amine > 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC300563-1G
APOSPC300563-1GEA
203
EUR
APOSPC300563-1G
N-Methyl-3-(trifluoromethyl)pyridin-2-amine > 95%
N-Methyl-3-(trifluoromethyl)pyridin-2-amine
Formel:
C₇H₇F₃N₂ Molecular Weight: 176,139 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11118894 CAS-Nummer: 1036584-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...