2-(Ethylamino)-5-(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
Synonyme:
N-[5-(Trifluoromethyl)pyridin-2-yl]ethylamine, N-Ethyl-5-(trifluoromethyl)pyridin-2-amine
PC300564-1G
PC300564-250MG
APOSPC300564-1GEA
186
EUR
APOSPC300564-1G
APOSPC300564-250MG
2-(Ethylamino)-5-(trifluoromethyl)pyridine 95%
2-(Ethylamino)-5-(trifluoromethyl)pyridine
Formel:
C₈H₉F₃N₂ Molecular Weight: 190.166 g/mol Schmelzpunkt: 74…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11118753 CAS-Nummer: 89810-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...