3-Chloro-N-propyl-5-(trifluoromethyl)-2-pyridinamine

Lieferant: Apollo Scientific

PC300567-1G
APOSPC300567-1GEA 241 EUR
APOSPC300567-1G
3-Chloro-N-propyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-propyl-5-(trifluoromethyl)-2-pyridinamine
Formel: C₉H₁₀ClF₃N₂
Molecular Weight: 238,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11119991
CAS-Nummer: 1041557-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR