2-Acetyl-6-fluoro-2H-indazole

Lieferant: Apollo Scientific
Achtung

PC300568-1G
APOSPC300568-1GEA 387 EUR
APOSPC300568-1G
2-Acetyl-6-fluoro-2H-indazole
2-Acetyl-6-fluoro-2H-indazole
Formel: C₉H₇FN₂O
Molecular Weight: 178,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15146800
CAS-Nummer: 1355171-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR