Ethyl-8-(trifluormethyl)imidazo[1,2-a]pyridin-2-carboxylat 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

PC300571-250MG
APOSPC300571-250MGEA 139 EUR
APOSPC300571-250MG
Ethyl-8-(trifluormethyl)imidazo[1,2-a]pyridin-2-carboxylat 95%
Ethyl-8-(trifluormethyl)imidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₉F₃N₂O₂
Molecular Weight: 258,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987556
CAS-Nummer: 1363404-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR