7-Fluoro-6-methoxy-1-methyl-1H-indazol-3-amine
Lieferant: Apollo Scientific
|
Achtung
|
PC300573-1G
APOSPC300573-1GEA
574
EUR
APOSPC300573-1G
7-Fluoro-6-methoxy-1-methyl-1H-indazol-3-amine
7-Fluoro-6-methoxy-1-methyl-1H-indazol-3-amine
Formel:
C₉H₁₀FN₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD16631266 CAS-Nummer: 1355171-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...