Ethyl 3-chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC300577-1G
APOSPC300577-1GEA
745
EUR
APOSPC300577-1G
Ethyl 3-chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-chloro-8-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₁H₈ClF₃N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD17168282 CAS-Nummer: 1355171-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...