3-Chloro-2-ethoxy-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC300579-1G PC300579-5G
APOSPC300579-1GEA 0 EUR
APOSPC300579-1G APOSPC300579-5G
3-Chloro-2-ethoxy-5-(trifluoromethyl)pyridine
3-Chloro-2-ethoxy-5-(trifluoromethyl)pyridine
Formel: C₈H₇ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD17168286
CAS-Nummer: 1379366-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR