Ethyl 3-bromo-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC300581-1G
APOSPC300581-1GEA 58 EUR
APOSPC300581-1G
Ethyl 3-bromo-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₁H₇BrClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17168288
CAS-Nummer: 1355170-29-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR