3-Chloro-4-fluorobenzhydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-4-fluorobenzohydrazide

PC300586-1G
APOSPC300586-1GEA 165 EUR
APOSPC300586-1G
3-Chloro-4-fluorobenzhydrazide
3-Chloro-4-fluorobenzhydrazide
Formel: C₇H₆ClFN₂O
Molecular Weight: 188.587 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037409
CAS-Nummer: 219783-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR