Ethyl 6-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC300587-1G
APOSPC300587-1GEA
232
EUR
APOSPC300587-1G
Ethyl 6-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 6-bromo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Formel:
C₁₁H₈BrF₃N₂O₂ Molecular Weight: 337,093 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18804152 CAS-Nummer: 1427460-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...