N-(2-Chloroethyl)-N'-[3-(difluoromethoxy)phenyl]urea

Lieferant: Apollo Scientific
Achtung

PC300588-1G
APOSPC300588-1GEA 192 EUR
APOSPC300588-1G
N-(2-Chloroethyl)-N'-[3-(difluoromethoxy)phenyl]urea
N-(2-Chloroethyl)-N'-[3-(difluoromethoxy)phenyl]urea
Formel: C₁₀H₁₁ClF₂N₂O₂
Molecular Weight: 264,656 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037410
CAS-Nummer: 1427460-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR