Methyl 3-fluoro-4-hydroxyphenylacetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-(2-methoxy-2-oxoethyl)phenol, 3-Fluoro-4-hydroxyphenylacetic acid methyl ester
PC300593-1G
APOSPC300593-1GEA
488
EUR
APOSPC300593-1G
Methyl 3-fluoro-4-hydroxyphenylacetate
Methyl 3-fluoro-4-hydroxyphenylacetate
Formel:
C₉H₉FO₃ Molecular Weight: 184,164 g/mol Siedepunkt: 90…93 °C (0,1 mm Hg) Schmelzpunkt: 32…36 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18399290 CAS-Nummer: 79280-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...