Ethyl 4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzoate

Lieferant: Apollo Scientific

PC300596-1G
APOSPC300596-1GEA 0 EUR
APOSPC300596-1G
Ethyl 4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Ethyl 4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Formel: C₁₅H₁₂F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18384921
CAS-Nummer: 1427460-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR