4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid

Lieferant: Apollo Scientific

PC300597-1G PC300597-5G
APOSPC300597-1GEA 0 EUR
APOSPC300597-1G APOSPC300597-5G
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Formel: C₁₃H₈F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08448326
CAS-Nummer: 869109-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR