1-[6-(Trifluoromethyl)pyridin-2-yl]homopiperazine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-[6-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepane

PC300598-1G
APOSPC300598-1GEA 248 EUR
APOSPC300598-1G
1-[6-(Trifluoromethyl)pyridin-2-yl]homopiperazine
1-[6-(Trifluoromethyl)pyridin-2-yl]homopiperazine
Formel: C₁₁H₁₄F₃N₃
Molecular Weight: 245,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756231
CAS-Nummer: 1227954-88-2
UN: 3267
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR