N-(2-Aminoethyl)-N-[6-(trifluoromethyl)pyridin-2-yl]amine

Lieferant: Apollo Scientific
Achtung

PC300599-1G
APOSPC300599-1GEA 227 EUR
APOSPC300599-1G
N-(2-Aminoethyl)-N-[6-(trifluoromethyl)pyridin-2-yl]amine
N-(2-Aminoethyl)-N-[6-(trifluoromethyl)pyridin-2-yl]amine
Formel: C₈H₁₀F₃N₃
Molecular Weight: 205,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00099693
CAS-Nummer: 1227955-19-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR