4-(4-Trifluoromethyl-2-pyrimidinyloxy)benzenesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC300600-1G
APOSPC300600-1GEA
282
EUR
APOSPC300600-1G
4-(4-Trifluoromethyl-2-pyrimidinyloxy)benzenesulphonyl chloride
4-(4-Trifluoromethyl-2-pyrimidinyloxy)benzenesulphonyl chloride
Formel:
C₁₁H₆ClF₃N₂O₃S Molecular Weight: 338,685 g/mol Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109532 CAS-Nummer: 1160058-86-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...