N,N-Dimethyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
Lieferant: Apollo Scientific
PC300601-1G
APOSPC300601-1GEA
361
EUR
APOSPC300601-1G
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
Formel:
C₁₃H₁₂F₃N₃O₃S Molecular Weight: 347,313 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11109533 CAS-Nummer: 1227954-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...