Ethyl 1-{[2-methyl-6-(trifluoromethyl)pyridin-3-yl]methyl}piperidine-3-carboxylate

Lieferant: Apollo Scientific

PC300603-1G
APOSPC300603-1GEA 371 EUR
APOSPC300603-1G
Ethyl 1-{[2-methyl-6-(trifluoromethyl)pyridin-3-yl]methyl}piperidine-3-carboxylate
Ethyl 1-{[2-methyl-6-(trifluoromethyl)pyridin-3-yl]methyl}piperidine-3-carboxylate
Formel: C₁₆H₂₁F₃N₂O₂
Molecular Weight: 330,345 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194592
CAS-Nummer: 1308645-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR