[2-(2-Fluorophenoxy)phenyl]methanol

Lieferant: Apollo Scientific

Synonyme: (2-(2-Fluorophenoxy)phenyl)methanol

PC300606-1G
APOSPC300606-1GEA 248 EUR
APOSPC300606-1G
[2-(2-Fluorophenoxy)phenyl]methanol
[2-(2-Fluorophenoxy)phenyl]methanol
Formel: C₁₃H₁₁FO₂
Molecular Weight: 218,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04108476
CAS-Nummer: 81890-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR