1-(Bromomethyl)-2-(2-fluorophenoxy)benzene

Lieferant: Apollo Scientific

PC300608-1G
APOSPC300608-1GEA 310 EUR
APOSPC300608-1G
1-(Bromomethyl)-2-(2-fluorophenoxy)benzene
1-(Bromomethyl)-2-(2-fluorophenoxy)benzene
Formel: C₁₃H₁₀BrFO
Molecular Weight: 281.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15146804
CAS-Nummer: 1355171-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR