4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl

Lieferant: Apollo Scientific
Gefahr

PC300618-1G
APOSPC300618-1GEA 361 EUR
APOSPC300618-1G
4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl
4-(Bromomethyl)-4'-fluoro-1,1'-biphenyl
Formel: C₁₃H₁₀BrF
Molecular Weight: 265,121 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15146810
CAS-Nummer: 147497-57-2
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR