2'-Fluoro-4-biphenylmethanol
Lieferant: Apollo Scientific
PC300620-1G
APOSPC300620-1GEA
296
EUR
APOSPC300620-1G
2'-Fluoro-4-biphenylmethanol
2'-Fluoro-4-biphenylmethanol
Formel:
C₁₃H₁₁FO Molecular Weight: 202,228 g/mol Schmelzpunkt: 57…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05980834 CAS-Nummer: 773872-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...