1-Nitro-4-phenoxy-2-(trifluoromethyl)benzene

Lieferant: Apollo Scientific

PC300622-1G
APOSPC300622-1GEA 186 EUR
APOSPC300622-1G
1-Nitro-4-phenoxy-2-(trifluoromethyl)benzene
1-Nitro-4-phenoxy-2-(trifluoromethyl)benzene
Formel: C₁₃H₈F₃NO₃
Molecular Weight: 283,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15146812
CAS-Nummer: 553666-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR