4-(4-Bromophenoxy)-1-nitro-2-(trifluoromethyl)benzene

Lieferant: Apollo Scientific

PC300624-1G
APOSPC300624-1GEA 183 EUR
APOSPC300624-1G
4-(4-Bromophenoxy)-1-nitro-2-(trifluoromethyl)benzene
4-(4-Bromophenoxy)-1-nitro-2-(trifluoromethyl)benzene
Formel: C₁₃H₇BrF₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD13677626
CAS-Nummer: 1156314-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR