4-(4-Bromophenoxy)-2-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

PC300625-1G
APOSPC300625-1GEA 241 EUR
APOSPC300625-1G
4-(4-Bromophenoxy)-2-(trifluoromethyl)aniline
4-(4-Bromophenoxy)-2-(trifluoromethyl)aniline
Formel: C₁₃H₉BrF₃NO
Molecular Weight: 332,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12444002
CAS-Nummer: 1040341-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR