1-[4-(Trifluoromethyl)pyridin-2-yl]homopiperazine

Lieferant: Apollo Scientific
Gefahr

PC300627-1G
APOSPC300627-1GEA 432 EUR
APOSPC300627-1G
1-[4-(Trifluoromethyl)pyridin-2-yl]homopiperazine
1-[4-(Trifluoromethyl)pyridin-2-yl]homopiperazine
Formel: C₁₁H₁₄F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09759191
CAS-Nummer: 1048921-30-7
UN: 3267
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR