5-(Trifluoromethyl)imidazo[1,2-a]pyrimidine

Lieferant: Apollo Scientific

PC300631-1G PC300631-10G PC300631-5G
APOSPC300631-1GEA 186 EUR
APOSPC300631-1G APOSPC300631-10G APOSPC300631-5G
5-(Trifluoromethyl)imidazo[1,2-a]pyrimidine
5-(Trifluoromethyl)imidazo[1,2-a]pyrimidine
Formel: C₇H₄F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09994617
CAS-Nummer: 1020033-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR