tert-Butyl 4-[2-(2-bromo-4-fluorophenoxy)ethyl]piperazine-1-carboxylate

Lieferant: Apollo Scientific

PC300641-1G PC300641-10G PC300641-5G
APOSPC300641-1GEA 288 EUR
APOSPC300641-1G APOSPC300641-10G APOSPC300641-5G
tert-Butyl 4-[2-(2-bromo-4-fluorophenoxy)ethyl]piperazine-1-carboxylate
tert-Butyl 4-[2-(2-bromo-4-fluorophenoxy)ethyl]piperazine-1-carboxylate
Formel: C₁₇H₂₄BrFN₂O₃
Molecular Weight: 403.286 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11615818
CAS-Nummer: 1227955-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR