2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde

Lieferant: Apollo Scientific

PC300648-1G PC300648-10G PC300648-5G
APOSPC300648-1GEA 193 EUR
APOSPC300648-1G APOSPC300648-10G APOSPC300648-5G
2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
Formel: C₁₂H₇F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11846261
CAS-Nummer: 1227954-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR