3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde

Lieferant: Apollo Scientific

PC300649-1G
APOSPC300649-1GEA 198 EUR
APOSPC300649-1G
3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
3-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
Formel: C₁₂H₇F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11505008
CAS-Nummer: 1216892-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR